Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTATCATGAAGGTTTGACTGATATT[A/G]CAAAGGATCCAGACTTTAATGAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609803 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKAR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKAR - ankyrin and armadillo repeat containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_144708.3 | 1190 | Missense Mutation | ACA,GCA | T,A 206 | NP_653309.3 | |
XM_011510673.2 | 1190 | Missense Mutation | ACA,GCA | T,A 206 | XP_011508975.1 | |
XM_011510674.1 | 1190 | Missense Mutation | ACA,GCA | T,A 206 | XP_011508976.1 | |
XM_011510675.2 | 1190 | Missense Mutation | ACA,GCA | T,A 206 | XP_011508977.1 | |
XM_011510676.2 | 1190 | Missense Mutation | ACA,GCA | T,A 206 | XP_011508978.1 | |
XM_011510677.2 | 1190 | Missense Mutation | ACA,GCA | T,A 206 | XP_011508979.1 | |
XM_011510679.2 | 1190 | Missense Mutation | ACA,GCA | T,A 206 | XP_011508981.1 | |
XM_011510680.1 | 1190 | Missense Mutation | ACA,GCA | T,A 206 | XP_011508982.1 | |
XM_011510681.1 | 1190 | Missense Mutation | ACA,GCA | T,A 206 | XP_011508983.1 | |
XM_011510682.2 | 1190 | Missense Mutation | ACA,GCA | T,A 206 | XP_011508984.1 | |
XM_011510685.2 | 1190 | Missense Mutation | ACA,GCA | T,A 54 | XP_011508987.1 | |
XM_011510686.2 | 1190 | Missense Mutation | ACA,GCA | T,A 206 | XP_011508988.1 | |
XM_011510687.2 | 1190 | UTR 5 | XP_011508989.1 | |||
XM_011510688.1 | 1190 | Missense Mutation | ACA,GCA | T,A 206 | XP_011508990.1 | |
XM_011510689.2 | 1190 | Missense Mutation | ACA,GCA | T,A 206 | XP_011508991.1 | |
XM_011510691.2 | 1190 | Intron | XP_011508993.1 | |||
XM_011510692.2 | 1190 | UTR 5 | XP_011508994.1 | |||
XM_011510693.2 | 1190 | UTR 5 | XP_011508995.1 | |||
XM_011510694.2 | 1190 | UTR 5 | XP_011508996.1 | |||
XM_017003413.1 | 1190 | Missense Mutation | ACA,GCA | T,A 206 | XP_016858902.1 | |
XM_017003414.1 | 1190 | Missense Mutation | ACA,GCA | T,A 206 | XP_016858903.1 | |
XM_017003415.1 | 1190 | UTR 5 | XP_016858904.1 | |||
XM_017003416.1 | 1190 | UTR 5 | XP_016858905.1 | |||
XM_017003417.1 | 1190 | Missense Mutation | ACA,GCA | T,A 206 | XP_016858906.1 | |
XM_017003418.1 | 1190 | UTR 5 | XP_016858907.1 | |||
XM_017003419.1 | 1190 | UTR 5 | XP_016858908.1 | |||
XM_017003420.1 | 1190 | UTR 5 | XP_016858909.1 |