Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGGACGGGAACGGGTCGCCCGTT[C/G]TGCCCGATAAGCGCAATGGTATCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616495 MIM: 612941 | ||||||||||||||||||||
Literature Links: |
ARL6IP6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARL6IP6 - ADP ribosylation factor like GTPase 6 interacting protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152522.5 | 979 | Silent Mutation | CTG,GTG | L,V 83 | NP_689735.1 | |
XM_006712326.3 | 979 | Intron | XP_006712389.1 |
PRPF40A - pre-mRNA processing factor 40 homolog A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017892.3 | 979 | Intron | NP_060362.3 | |||
XM_005246676.4 | 979 | Intron | XP_005246733.3 | |||
XM_005246677.4 | 979 | Intron | XP_005246734.2 | |||
XM_011511446.2 | 979 | Intron | XP_011509748.1 | |||
XM_011511447.2 | 979 | Intron | XP_011509749.1 | |||
XM_011511448.2 | 979 | Intron | XP_011509750.1 | |||
XM_011511449.2 | 979 | Intron | XP_011509751.1 | |||
XM_017004453.1 | 979 | Intron | XP_016859942.1 | |||
XM_017004454.1 | 979 | Intron | XP_016859943.1 | |||
XM_017004455.1 | 979 | Intron | XP_016859944.1 | |||
XM_017004456.1 | 979 | Intron | XP_016859945.1 | |||
XM_017004457.1 | 979 | Intron | XP_016859946.1 | |||
XM_017004458.1 | 979 | Intron | XP_016859947.1 | |||
XM_017004459.1 | 979 | Intron | XP_016859948.1 | |||
XM_017004460.1 | 979 | Intron | XP_016859949.1 | |||
XM_017004461.1 | 979 | Intron | XP_016859950.1 |