Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACTGTGGAGAGTTCGCTGAGCAT[A/G]TCCTTGTAGTGGGACCTCAGTTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611518 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOCK10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DOCK10 - dedicator of cytokinesis 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290263.1 | 6723 | Silent Mutation | NP_001277192.1 | |||
NM_014689.2 | 6723 | Silent Mutation | NP_055504.2 | |||
XM_005246673.4 | 6723 | Silent Mutation | XP_005246730.1 | |||
XM_006712617.3 | 6723 | Silent Mutation | XP_006712680.1 | |||
XM_017004441.1 | 6723 | Silent Mutation | XP_016859930.1 | |||
XM_017004442.1 | 6723 | Silent Mutation | XP_016859931.1 | |||
XM_017004443.1 | 6723 | Silent Mutation | XP_016859932.1 | |||
XM_017004444.1 | 6723 | Missense Mutation | XP_016859933.1 | |||
XM_017004445.1 | 6723 | Silent Mutation | XP_016859934.1 | |||
XM_017004446.1 | 6723 | Silent Mutation | XP_016859935.1 | |||
XM_017004447.1 | 6723 | Silent Mutation | XP_016859936.1 |