Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGACAACAGCAGTGGACATCTA[C/T]TCCTTTGGCATGTGTGCACTGGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607386 MIM: 606010 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFT172 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IFT172 - intraflagellar transport 172 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KRTCAP3 - keratinocyte associated protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NRBP1 - nuclear receptor binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321357.1 | 1612 | Silent Mutation | TAC,TAT | Y,Y 260 | NP_001308286.1 | |
NM_001321358.1 | 1612 | Silent Mutation | TAC,TAT | Y,Y 268 | NP_001308287.1 | |
NM_001321359.1 | 1612 | Silent Mutation | TAC,TAT | Y,Y 268 | NP_001308288.1 | |
NM_001321361.1 | 1612 | Silent Mutation | TAC,TAT | Y,Y 268 | NP_001308290.1 | |
NM_001321362.1 | 1612 | Silent Mutation | TAC,TAT | Y,Y 259 | NP_001308291.1 | |
NM_001321363.1 | 1612 | Silent Mutation | TAC,TAT | Y,Y 228 | NP_001308292.1 | |
NM_013392.3 | 1612 | Silent Mutation | TAC,TAT | Y,Y 260 | NP_037524.1 |