Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCAGAAGAACCCACTGTTGGTGG[G/T]TCTATATTTAATAAATGTTGTGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FARP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FARP2 - FERM, ARH/RhoGEF and pleckstrin domain protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282983.1 | Intron | NP_001269912.1 | ||||
NM_001282984.1 | Intron | NP_001269913.1 | ||||
NM_014808.3 | Intron | NP_055623.1 | ||||
XM_005247050.3 | Intron | XP_005247107.1 | ||||
XM_011512233.1 | Intron | XP_011510535.1 | ||||
XM_011512234.1 | Intron | XP_011510536.1 | ||||
XM_011512235.1 | Intron | XP_011510537.1 | ||||
XM_011512236.1 | Intron | XP_011510538.1 | ||||
XM_017005416.1 | Intron | XP_016860905.1 | ||||
XM_017005417.1 | Intron | XP_016860906.1 | ||||
XM_017005418.1 | Intron | XP_016860907.1 | ||||
XM_017005419.1 | Intron | XP_016860908.1 | ||||
XM_017005420.1 | Intron | XP_016860909.1 |