Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCGTGCATGCCTCTAGCCGAGGC[A/T]TCTTGGACAAAATGCTCTGCGGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CATIP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CATIP - ciliogenesis associated TTC17 interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320865.1 | 480 | Missense Mutation | ATC,TTC | I,F 106 | NP_001307794.1 | |
NM_198559.1 | 480 | Missense Mutation | ATC,TTC | I,F 95 | NP_940961.1 | |
XM_005246539.4 | 480 | Missense Mutation | ATC,TTC | I,F 64 | XP_005246596.1 | |
XM_005246541.4 | 480 | Intron | XP_005246598.1 | |||
XM_011511148.2 | 480 | Missense Mutation | ATC,TTC | I,F 106 | XP_011509450.1 | |
XM_011511149.2 | 480 | UTR 5 | XP_011509451.1 | |||
XM_011511150.2 | 480 | Intron | XP_011509452.1 | |||
XM_017004053.1 | 480 | UTR 5 | XP_016859542.1 |
CATIP-AS1 - CATIP antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CATIP-AS2 - CATIP antisense RNA 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |