Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCCAGTGGCTTCTCGGTCTTCA[C/T]CACCTTGCCCGACTTGCTCTTCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 188860 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAL - mal, T-cell differentiation protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002371.3 | 197 | Missense Mutation | ACC,ATC | T,I 20 | NP_002362.1 | |
NM_022438.2 | 197 | Missense Mutation | ACC,ATC | T,I 20 | NP_071883.1 | |
NM_022439.2 | 197 | Missense Mutation | ACC,ATC | T,I 20 | NP_071884.1 | |
NM_022440.2 | 197 | Missense Mutation | ACC,ATC | T,I 20 | NP_071885.1 |