Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGGGCAGGAGGGGCTGCGGGGCC[A/G]GAGGCACACCAGGCATATAGGATAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612195 MIM: 606395 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABHD1 - abhydrolase domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PREB - prolactin regulatory element binding | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRR30 - proline rich 30 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178553.3 | 1349 | Missense Mutation | CGG,TGG | R,W 257 | NP_848648.2 |