Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACATTTTGTATTTCACTGGTTCAG[C/T]TTAACAGCAAATATTTGAGATCTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601826 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DGKD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DGKD - diacylglycerol kinase delta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003648.2 | Intron | NP_003639.2 | ||||
NM_152879.2 | Intron | NP_690618.2 | ||||
XM_005246107.3 | Intron | XP_005246164.1 | ||||
XM_005246108.1 | Intron | XP_005246165.1 | ||||
XM_005246110.3 | Intron | XP_005246167.1 | ||||
XM_011512028.2 | Intron | XP_011510330.1 | ||||
XM_011512029.2 | Intron | XP_011510331.1 | ||||
XM_011512030.2 | Intron | XP_011510332.1 | ||||
XM_011512031.2 | Intron | XP_011510333.1 | ||||
XM_011512032.2 | Intron | XP_011510334.1 | ||||
XM_011512033.2 | Intron | XP_011510335.1 | ||||
XM_011512035.1 | Intron | XP_011510337.1 | ||||
XM_011512037.2 | Intron | XP_011510339.1 | ||||
XM_011512039.2 | Intron | XP_011510341.1 | ||||
XM_011512040.2 | Intron | XP_011510342.1 | ||||
XM_017005138.1 | Intron | XP_016860627.1 | ||||
XM_017005139.1 | Intron | XP_016860628.1 | ||||
XM_017005140.1 | Intron | XP_016860629.1 |