Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTTAGGACCCTGTGGATTTAGAC[A/G]TATACCAAAGCTCCCACATGGTCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TEX37 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TEX37 - testis expressed 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152670.2 | 163 | Missense Mutation | ATA,GTA | I,V 16 | NP_689883.1 | |
XM_005264188.3 | 163 | Missense Mutation | ATA,GTA | I,V 16 | XP_005264245.3 | |
XM_011532691.1 | 163 | Missense Mutation | ATA,GTA | I,V 16 | XP_011530993.1 | |
XM_011532692.1 | 163 | Missense Mutation | ATA,GTA | I,V 16 | XP_011530994.1 |