Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCCGGCCCAGCGTCTACCGCAAC[A/G]TGGCGCGTCAGCTGCACATCTCCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605404 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BOK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BOK - BOK, BCL2 family apoptosis regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032515.4 | 358 | Missense Mutation | ATG,GTG | M,V 88 | NP_115904.1 | |
XM_011511696.2 | 358 | Missense Mutation | ATG,GTG | M,V 88 | XP_011509998.1 | |
XM_011511697.2 | 358 | Missense Mutation | ATG,GTG | M,V 88 | XP_011509999.1 | |
XM_017004775.1 | 358 | Missense Mutation | ATG,GTG | M,V 10 | XP_016860264.1 | |
XM_017004776.1 | 358 | Missense Mutation | ATG,GTG | M,V 10 | XP_016860265.1 |
BOK-AS1 - BOK antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |