Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTCTCCTTTAGGGACTTGGAAAC[A/G]GAAGGTAGTGCAGTTCTTTCTGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609207 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MREG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MREG - melanoregulin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018000.2 | 925 | Missense Mutation | CCG,CTG | P,L 210 | NP_060470.2 | |
XM_011511468.2 | 925 | Missense Mutation | CCG,CTG | P,L 260 | XP_011509770.1 | |
XM_017004475.1 | 925 | Intron | XP_016859964.1 |