Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCTGAAGACATCTGACCTGGACAG[C/G]CTGCCATCTCTTATGATTATTTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600620 MIM: 616234 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FKBP1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FKBP1B - FK506 binding protein 1B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MFSD2B - major facilitator superfamily domain containing 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDCP - WD repeat and coiled coil containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142319.1 | 2159 | UTR 3 | NP_001135791.1 | |||
NM_025203.2 | 2159 | Missense Mutation | AGC,AGG | S,R 676 | NP_079479.1 | |
XM_017005029.1 | 2159 | Intron | XP_016860518.1 |