Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCTCCCCATAGGTGCCCCGCAC[C/T]GTCCTCTGGCTGACCATCGAGCTAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600266 | ||||||||||||||||||||
Literature Links: |
SLC11A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SLC11A1 - solute carrier family 11 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000578.3 | 1082 | Silent Mutation | ACC,ACT | T,T 135 | NP_000569.3 | |
XM_005246793.3 | 1082 | Silent Mutation | ACC,ACT | T,T 68 | XP_005246850.1 | |
XM_005246794.3 | 1082 | Silent Mutation | ACC,ACT | T,T 17 | XP_005246851.1 | |
XM_006712709.3 | 1082 | Silent Mutation | ACC,ACT | T,T 17 | XP_006712772.1 | |
XM_006712710.3 | 1082 | Silent Mutation | ACC,ACT | T,T 17 | XP_006712773.1 | |
XM_006712711.3 | 1082 | UTR 5 | XP_006712774.1 | |||
XM_011511684.2 | 1082 | Missense Mutation | CGT,TGT | R,C 15 | XP_011509986.1 | |
XM_011511685.2 | 1082 | Missense Mutation | CGT,TGT | R,C 15 | XP_011509987.1 | |
XM_017004765.1 | 1082 | Silent Mutation | ACC,ACT | T,T 94 | XP_016860254.1 | |
XM_017004766.1 | 1082 | Silent Mutation | ACC,ACT | T,T 68 | XP_016860255.1 | |
XM_017004767.1 | 1082 | Silent Mutation | ACC,ACT | T,T 135 | XP_016860256.1 |