Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGGAGAAAAGTAAACGGGCACC[A/G]ACTCATCAACGATTTCCTTTCCTCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608357 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SULT1C4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SULT1C4 - sulfotransferase family 1C member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321770.1 | 670 | Silent Mutation | CCA,CCG | P,P 81 | NP_001308699.1 | |
NM_006588.3 | 670 | Silent Mutation | CCA,CCG | P,P 81 | NP_006579.2 | |
XM_017003807.1 | 670 | UTR 5 | XP_016859296.1 |