Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606715 MIM: 610405 MIM: 610991 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASIC4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASIC4 - acid sensing ion channel subunit family member 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CHPF - chondroitin polymerizing factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3132 - microRNA 3132 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OBSL1 - obscurin like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM198 - transmembrane protein 198 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005209.2 | 632 | Missense Mutation | GGG,GTG | G,V 74 | NP_001005209.1 | |
NM_001303098.1 | 632 | Missense Mutation | GGG,GTG | G,V 74 | NP_001290027.1 | |
XM_017003367.1 | 632 | Missense Mutation | GGG,GTG | G,V 74 | XP_016858856.1 |