Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGCAACGCCGCGGGGGAGGCCGC[C/G]CCGGGCCCGCGAGTGCTGGGCTTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607804 MIM: 607805 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CNNM3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CNNM3 - cyclin and CBS domain divalent metal cation transport mediator 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017623.4 | 108 | Silent Mutation | GCC,GCG | A,A 28 | NP_060093.3 | |
NM_199078.2 | 108 | Silent Mutation | GCC,GCG | A,A 28 | NP_951060.1 | |
XM_011510957.2 | 108 | Silent Mutation | GCC,GCG | A,A 28 | XP_011509259.1 | |
XM_017003800.1 | 108 | Silent Mutation | GCC,GCG | A,A 28 | XP_016859289.1 |
CNNM4 - cyclin and CBS domain divalent metal cation transport mediator 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |