Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGACCTCCCCTTCCTTTCACATG[A/G]GCTGGTCTGACAATATTTATTGGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602507 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105374845 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105374845 - uncharacterized LOC105374845 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RNF103 - ring finger protein 103 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198951.1 | 1762 | Missense Mutation | CCA,TCA | P,S 583 | NP_001185880.1 | |
NM_001198952.1 | 1762 | Intron | NP_001185881.1 | |||
NM_005667.3 | 1762 | Missense Mutation | CCA,TCA | P,S 587 | NP_005658.1 |
RNF103-CHMP3 - RNF103-CHMP3 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198954.1 | 1762 | Intron | NP_001185883.1 |