Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTATGTAAGTGGCATCCAGAGGA[A/G]AGTAGGTGATACAATCCATCATGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120070 MIM: 120131 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COL4A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COL4A3 - collagen type IV alpha 3 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000091.4 | Intron | NP_000082.2 | ||||
XM_005246277.3 | Intron | XP_005246334.1 | ||||
XM_005246280.3 | Intron | XP_005246337.1 | ||||
XM_006712245.3 | Intron | XP_006712308.1 | ||||
XM_011510555.1 | Intron | XP_011508857.1 | ||||
XM_011510556.2 | Intron | XP_011508858.1 | ||||
XM_017003295.1 | Intron | XP_016858784.1 |
COL4A4 - collagen type IV alpha 4 chain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |