Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGGGGCGGCCTGGGCTGGTGTATC[A/G]TGGTCCTGGATGTACTGCAAGATGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609359 | ||||||||||||||||||||
Literature Links: |
HS1BP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HS1BP3 - HCLS1 binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022460.3 | 1184 | Silent Mutation | CAC,CAT | H,H 381 | NP_071905.3 | |
XM_017004696.1 | 1184 | Intron | XP_016860185.1 | |||
XM_017004697.1 | 1184 | Intron | XP_016860186.1 | |||
XM_017004698.1 | 1184 | Intron | XP_016860187.1 | |||
XM_017004699.1 | 1184 | Intron | XP_016860188.1 | |||
XM_017004700.1 | 1184 | Silent Mutation | CAC,CAT | H,H 348 | XP_016860189.1 | |
XM_017004701.1 | 1184 | Intron | XP_016860190.1 |