Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCAGGCACTGAAAGACTCTCAGA[G/T]GCCACTGCTGGGGACAAAATCCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604285 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGXT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGXT - alanine-glyoxylate aminotransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C2orf54 - chromosome 2 open reading frame 54 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085437.2 | 1647 | UTR 3 | NP_001078906.2 | |||
NM_001282921.1 | 1647 | UTR 3 | NP_001269850.1 | |||
NM_024861.3 | 1647 | UTR 3 | NP_079137.3 | |||
XM_011511877.1 | 1647 | UTR 3 | XP_011510179.1 |