Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACGGGGTCACCTTCTTGTAGGTGA[C/T]GTGAAGATGCTGAGTCATTGGCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612533 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
THAP4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
THAP4 - THAP domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164356.1 | 2001 | Missense Mutation | NP_001157828.1 | |||
NM_015963.5 | 2001 | Missense Mutation | NP_057047.4 | |||
XM_005247016.4 | 2001 | Missense Mutation | XP_005247073.3 | |||
XM_011511291.2 | 2001 | Missense Mutation | XP_011509593.1 | |||
XM_017004256.1 | 2001 | Missense Mutation | XP_016859745.1 |