Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCCGGAAGGGCTTGATTGACGTG[A/G]AGATGTACAACTCGACTCAAGGGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 137570 | ||||||||||||||||||||
Literature Links: |
FLJ42351 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FLJ42351 - uncharacterized LOC400999 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC20A1 - solute carrier family 20 member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005415.4 | 787 | Missense Mutation | AAG,GAG | K,E 93 | NP_005406.3 | |
XM_017004768.1 | 787 | UTR 5 | XP_016860257.1 | |||
XM_017004769.1 | 787 | UTR 5 | XP_016860258.1 | |||
XM_017004770.1 | 787 | UTR 5 | XP_016860259.1 | |||
XM_017004771.1 | 787 | UTR 5 | XP_016860260.1 |