Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTATGACCAGGTTGCCAGTGTAC[C/T]GGTCTAGCAACATAGGGAAATGATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614029 | ||||||||||||||||||||
Literature Links: |
SPDYA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SPDYA - speedy/RINGO cell cycle regulator family member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008779.1 | 1076 | Intron | NP_001008779.1 | |||
NM_001142634.1 | 1076 | UTR 3 | NP_001136106.1 | |||
NM_182756.3 | 1076 | UTR 3 | NP_877433.2 |
TRMT61B - tRNA methyltransferase 61B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017910.3 | 1076 | Silent Mutation | CCA,CCG | P,P 455 | NP_060380.3 | |
XM_005264393.2 | 1076 | Silent Mutation | CCA,CCG | P,P 446 | XP_005264450.1 | |
XM_006712037.2 | 1076 | Silent Mutation | CCA,CCG | P,P 357 | XP_006712100.1 | |
XM_017004401.1 | 1076 | Silent Mutation | CCA,CCG | P,P 348 | XP_016859890.1 | |
XM_017004402.1 | 1076 | Intron | XP_016859891.1 | |||
XM_017004403.1 | 1076 | Silent Mutation | CCA,CCG | P,P 219 | XP_016859892.1 |