Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTAATAGCAATCAAAACAGAGTA[A/G]GCGGAAGGTCTTTTTTGTAAGGGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610059 MIM: 611132 | ||||||||||||||||||||
Literature Links: |
MRPL33 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MRPL33 - mitochondrial ribosomal protein L33 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RBKS - ribokinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287580.1 | 2126 | Missense Mutation | CCT,CTT | P,L 252 | NP_001274509.1 | |
NM_022128.2 | 2126 | Missense Mutation | CCT,CTT | P,L 319 | NP_071411.1 |