Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAACCATCATCCAGGGAGACAAC[A/G]AACTCTCCTCTTTGAAGCTGCACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
DHX57 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DHX57 - DEAH-box helicase 57 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011533154.2 | 4069 | Silent Mutation | TTC,TTT | F,F 1322 | XP_011531456.1 | |
XM_011533155.2 | 4069 | Silent Mutation | TTC,TTT | F,F 1322 | XP_011531457.1 | |
XM_011533156.2 | 4069 | Silent Mutation | TTC,TTT | F,F 1322 | XP_011531458.1 | |
XM_011533157.1 | 4069 | Intron | XP_011531459.1 |