Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGTGGTTTATCATGAGCTGATCCA[A/G]AGGATGTACACTCGCCTGGAGCCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CNPPD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CNPPD1 - cyclin Pas1/PHO80 domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM134A - family with sequence similarity 134 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321109.1 | 853 | Silent Mutation | CAA,CAG | Q,Q 23 | NP_001308038.1 | |
NM_001321110.1 | 853 | Silent Mutation | CAA,CAG | Q,Q 23 | NP_001308039.1 | |
NM_024293.5 | 853 | Silent Mutation | CAA,CAG | Q,Q 230 | NP_077269.3 | |
XM_005246848.2 | 853 | Silent Mutation | CAA,CAG | Q,Q 230 | XP_005246905.1 |