Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGACGATGGTCTCAAAGGGTCCC[A/G]GACGGCAGCCACAGCGTCCGACTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
UBE2F PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UBE2F - ubiquitin conjugating enzyme E2 F (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278305.1 | 257 | Missense Mutation | CAG,CGG | Q,R 18 | NP_001265234.1 | |
NM_001278306.1 | 257 | Intron | NP_001265235.1 | |||
NM_001278307.1 | 257 | Missense Mutation | CAG,CGG | Q,R 18 | NP_001265236.1 | |
NM_001278308.1 | 257 | Missense Mutation | CAG,CGG | Q,R 18 | NP_001265237.1 | |
NM_080678.2 | 257 | Missense Mutation | CAG,CGG | Q,R 18 | NP_542409.1 |
UBE2F-SCLY - UBE2F-SCLY readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |