Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTAGTAAACAATTGTTAAAGTCTG[A/G]TGTTAACTACCAGTGTTTATTTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609803 | ||||||||||||||||||||
Literature Links: |
ANKAR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKAR - ankyrin and armadillo repeat containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ASNSD1 - asparagine synthetase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019048.2 | 1075 | Missense Mutation | GAT,GGT | D,G 42 | NP_061921.1 | |
XM_017004381.1 | 1075 | Missense Mutation | GAT,GGT | D,G 42 | XP_016859870.1 | |
XM_017004382.1 | 1075 | Missense Mutation | GAT,GGT | D,G 42 | XP_016859871.1 |