Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGCAAACAAGCATGAAGACTCC[A/G]AGGAGTATGTGGTCCATCACCATTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 100690 | ||||||||||||||||||||
Literature Links: |
CHRNA1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHRNA1 - cholinergic receptor nicotinic alpha 1 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000079.3 | 1371 | Silent Mutation | CTC,CTT | L,L 431 | NP_000070.1 | |
NM_001039523.2 | 1371 | Silent Mutation | CTC,CTT | L,L 456 | NP_001034612.1 | |
XM_017003256.1 | 1371 | Silent Mutation | CTC,CTT | L,L 463 | XP_016858745.1 | |
XM_017003257.1 | 1371 | Silent Mutation | CTC,CTT | L,L 438 | XP_016858746.1 |