Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGCTGTGAATGATTGTATGCTGA[A/G]GTCAGTTCAGAGAAAAGGTAATGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610587 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGAP25 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGAP25 - Rho GTPase activating protein 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007231.2 | Intron | NP_001007232.2 | ||||
NM_001166276.1 | Intron | NP_001159748.1 | ||||
NM_001166277.1 | Intron | NP_001159749.1 | ||||
NM_014882.2 | Intron | NP_055697.1 | ||||
XM_005264675.2 | Intron | XP_005264732.1 | ||||
XM_005264676.2 | Intron | XP_005264733.1 | ||||
XM_011533206.2 | Intron | XP_011531508.1 | ||||
XM_011533207.2 | Intron | XP_011531509.1 | ||||
XM_011533208.2 | Intron | XP_011531510.1 | ||||
XM_011533209.2 | Intron | XP_011531511.1 | ||||
XM_011533210.2 | Intron | XP_011531512.1 | ||||
XM_017005423.1 | Intron | XP_016860912.1 | ||||
XM_017005424.1 | Intron | XP_016860913.1 | ||||
XM_017005425.1 | Intron | XP_016860914.1 | ||||
XM_017005426.1 | Intron | XP_016860915.1 | ||||
XM_017005427.1 | Intron | XP_016860916.1 | ||||
XM_017005428.1 | Intron | XP_016860917.1 |