Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTGTCAACTGCGTTGTCAGAGAT[G/T]ATCACCGCCCGCCCGCCGTGCTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605140 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCT7 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCT7 - chaperonin containing TCP1 subunit 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRADC1 - protease associated domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032319.1 | 407 | Silent Mutation | ATA,ATC | I,I 113 | NP_115695.1 |
SMYD5 - SMYD family member 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |