Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGCCCGGCCCCACTGCCGCCCTC[C/T]ACGGTGCCATTCACAGTTGCTGGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601248 | ||||||||||||||||||||
Literature Links: |
BIN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BIN1 - bridging integrator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320632.1 | 2020 | Silent Mutation | GTA,GTG | V,V 337 | NP_001307561.1 | |
NM_001320633.1 | 2020 | Silent Mutation | GTA,GTG | V,V 383 | NP_001307562.1 | |
NM_001320634.1 | 2020 | Silent Mutation | GTA,GTG | V,V 298 | NP_001307563.1 | |
NM_001320640.1 | 2020 | Silent Mutation | GTA,GTG | V,V 426 | NP_001307569.1 | |
NM_001320641.1 | 2020 | Silent Mutation | GTA,GTG | V,V 475 | NP_001307570.1 | |
NM_001320642.1 | 2020 | Silent Mutation | GTA,GTG | V,V 479 | NP_001307571.1 | |
NM_004305.3 | 2020 | Silent Mutation | GTA,GTG | V,V 367 | NP_004296.1 | |
NM_139343.2 | 2020 | Silent Mutation | GTA,GTG | V,V 506 | NP_647593.1 | |
NM_139344.2 | 2020 | Silent Mutation | GTA,GTG | V,V 463 | NP_647594.1 | |
NM_139345.2 | 2020 | Silent Mutation | GTA,GTG | V,V 419 | NP_647595.1 | |
NM_139346.2 | 2020 | Silent Mutation | GTA,GTG | V,V 410 | NP_647596.1 | |
NM_139347.2 | 2020 | Silent Mutation | GTA,GTG | V,V 431 | NP_647597.1 | |
NM_139348.2 | 2020 | Silent Mutation | GTA,GTG | V,V 395 | NP_647598.1 | |
NM_139349.2 | 2020 | Silent Mutation | GTA,GTG | V,V 388 | NP_647599.1 | |
NM_139350.2 | 2020 | Silent Mutation | GTA,GTG | V,V 352 | NP_647600.1 | |
NM_139351.2 | 2020 | Silent Mutation | GTA,GTG | V,V 322 | NP_647601.1 | |
XM_005263642.3 | 2020 | Silent Mutation | GTA,GTG | V,V 521 | XP_005263699.1 | |
XM_005263643.3 | 2020 | Silent Mutation | GTA,GTG | V,V 490 | XP_005263700.1 | |
XM_005263645.2 | 2020 | Silent Mutation | GTA,GTG | V,V 466 | XP_005263702.1 | |
XM_006712425.3 | 2020 | Silent Mutation | GTA,GTG | V,V 477 | XP_006712488.1 | |
XM_006712426.3 | 2020 | Silent Mutation | GTA,GTG | V,V 477 | XP_006712489.1 | |
XM_006712427.2 | 2020 | Silent Mutation | GTA,GTG | V,V 451 | XP_006712490.1 | |
XM_006712428.3 | 2020 | Silent Mutation | GTA,GTG | V,V 441 | XP_006712491.1 | |
XM_006712429.3 | 2020 | Silent Mutation | GTA,GTG | V,V 434 | XP_006712492.1 | |
XM_006712431.3 | 2020 | Silent Mutation | GTA,GTG | V,V 411 | XP_006712494.1 | |
XM_006712432.3 | 2020 | Silent Mutation | GTA,GTG | V,V 398 | XP_006712495.1 | |
XM_006712433.3 | 2020 | Silent Mutation | GTA,GTG | V,V 368 | XP_006712496.1 | |
XM_006712434.3 | 2020 | Silent Mutation | GTA,GTG | V,V 353 | XP_006712497.1 | |
XM_011510975.2 | 2020 | Silent Mutation | GTA,GTG | V,V 494 | XP_011509277.1 | |
XM_017003819.1 | 2020 | Silent Mutation | GTA,GTG | V,V 497 | XP_016859308.1 | |
XM_017003820.1 | 2020 | Silent Mutation | GTA,GTG | V,V 431 | XP_016859309.1 | |
XM_017003821.1 | 2020 | Silent Mutation | GTA,GTG | V,V 396 | XP_016859310.1 | |
XM_017003822.1 | 2020 | Silent Mutation | GTA,GTG | V,V 365 | XP_016859311.1 | |
XM_017003823.1 | 2020 | Silent Mutation | GTA,GTG | V,V 364 | XP_016859312.1 | |
XM_017003824.1 | 2020 | Silent Mutation | GTA,GTG | V,V 341 | XP_016859313.1 | |
XM_017003825.1 | 2020 | Silent Mutation | GTA,GTG | V,V 329 | XP_016859314.1 | |
XM_017003826.1 | 2020 | Silent Mutation | GTA,GTG | V,V 328 | XP_016859315.1 | |
XM_017003827.1 | 2020 | Intron | XP_016859316.1 | |||
XM_017003828.1 | 2020 | Intron | XP_016859317.1 |