Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATCTTCAGAAAAGACTTGCTCCTG[A/G]TAACTGTCTAGCCATCCTAAGATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603650 MIM: 607701 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BBS5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BBS5 - Bardet-Biedl syndrome 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLHL41 - kelch like family member 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006063.2 | 475 | Missense Mutation | GAT,GGT | D,G 133 | NP_006054.2 |