Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCAGCATACTTCAGCAAAGATG[C/T]GTAACGCTTTAGAAACAGATCATCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605460 MIM: 607544 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCG8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABCG8 - ATP binding cassette subfamily G member 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRPPRC - leucine rich pentatricopeptide repeat containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_133259.3 | 4045 | Missense Mutation | ACA,GCA | T,A 1360 | NP_573566.2 | |
XM_006711915.2 | 4045 | Missense Mutation | ACA,GCA | T,A 1334 | XP_006711978.1 | |
XM_006711916.3 | 4045 | Intron | XP_006711979.1 | |||
XM_011532473.1 | 4045 | Missense Mutation | ACA,GCA | T,A 1335 | XP_011530775.1 | |
XM_017003117.1 | 4045 | Missense Mutation | ACA,GCA | T,A 1309 | XP_016858606.1 |