Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGAAGGTGCAGAGCGCTGAGCT[G/T]CCTTTCTGCGGCTACCACTTTGAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613817 | ||||||||||||||||||||
Literature Links: |
KCTD18 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KCTD18 - potassium channel tetramerization domain containing 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321547.1 | 1507 | Missense Mutation | GAA,GCA | E,A 322 | NP_001308476.1 | |
NM_001321548.1 | 1507 | Missense Mutation | GAA,GCA | E,A 113 | NP_001308477.1 | |
NM_001321550.1 | 1507 | Missense Mutation | GAA,GCA | E,A 113 | NP_001308479.1 | |
NM_152387.3 | 1507 | Missense Mutation | GAA,GCA | E,A 322 | NP_689600.2 |
SPATS2L - spermatogenesis associated serine rich 2 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |