Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTGCTGCCACAAGTATCTGAAA[C/T]GGATCAACAATCTACCAAAGCAAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608680 | ||||||||||||||||||||
Literature Links: |
TPRKB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TPRKB - TP53RK binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005264343.3 | 624 | Intron | XP_005264400.1 | |||
XM_006712026.3 | 624 | Silent Mutation | CCA,CCG | P,P 90 | XP_006712089.1 | |
XM_006712027.3 | 624 | Silent Mutation | CCA,CCG | P,P 90 | XP_006712090.1 | |
XM_011532877.2 | 624 | Silent Mutation | CCA,CCG | P,P 90 | XP_011531179.1 | |
XM_011532878.2 | 624 | Intron | XP_011531180.1 | |||
XM_011532879.2 | 624 | Silent Mutation | CCA,CCG | P,P 18 | XP_011531181.1 | |
XM_011532880.2 | 624 | Silent Mutation | CCA,CCG | P,P 18 | XP_011531182.1 | |
XM_017004232.1 | 624 | Silent Mutation | CCA,CCG | P,P 90 | XP_016859721.1 | |
XM_017004233.1 | 624 | Silent Mutation | CCA,CCG | P,P 90 | XP_016859722.1 | |
XM_017004234.1 | 624 | Silent Mutation | CCA,CCG | P,P 51 | XP_016859723.1 | |
XM_017004235.1 | 624 | Silent Mutation | CCA,CCG | P,P 51 | XP_016859724.1 |