Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCAGGTCAAGTCCAAATGTAGAT[C/T]CTTGGGAGACTGCTGCTACAGGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604930 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NCK2 - NCK adaptor protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004720.2 | Intron | NP_001004720.1 | ||||
NM_001004722.3 | Intron | NP_001004722.1 | ||||
NM_003581.4 | Intron | NP_003572.2 | ||||
XM_006712797.3 | Intron | XP_006712860.1 | ||||
XM_011511991.2 | Intron | XP_011510293.1 | ||||
XM_011511992.2 | Intron | XP_011510294.2 | ||||
XM_017005103.1 | Intron | XP_016860592.1 | ||||
XM_017005104.1 | Intron | XP_016860593.1 | ||||
XM_017005105.1 | Intron | XP_016860594.1 |