Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGCAGGGCACTCTTTATCATTTC[A/G]AGGCACACTCTGTTTTATCCCAAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611655 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PGAP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PGAP1 - post-GPI attachment to proteins 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321099.1 | 9495 | UTR 3 | NP_001308028.1 | |||
NM_001321100.1 | 9495 | UTR 3 | NP_001308029.1 | |||
NM_024989.3 | 9495 | UTR 3 | NP_079265.2 | |||
XM_017004992.1 | 9495 | UTR 3 | XP_016860481.1 | |||
XM_017004993.1 | 9495 | UTR 3 | XP_016860482.1 | |||
XM_017004994.1 | 9495 | UTR 3 | XP_016860483.1 |