Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGGAAGATGGCGGAACAGGCTAC[C/G]AAGTCCGTGCTGTTTGTGTGTCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 171500 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACP1 - acid phosphatase 1, soluble | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040649.2 | 114 | Silent Mutation | ACC,ACG | T,T 6 | NP_001035739.1 | |
NM_004300.3 | 114 | Intron | NP_004291.1 | |||
NM_007099.3 | 114 | Intron | NP_009030.1 |
SH3YL1 - SH3 and SYLF domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159597.2 | 114 | Intron | NP_001153069.1 | |||
NM_001282682.1 | 114 | Intron | NP_001269611.1 | |||
NM_001282687.1 | 114 | Intron | NP_001269616.1 | |||
NM_015677.3 | 114 | Intron | NP_056492.2 |