Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCATCATCATCATCAATTTCTTCA[C/T]CCCAAACCTTCTGCTCCTCATCATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610072 MIM: 615129 | ||||||||||||||||||||
Literature Links: |
ERMN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ERMN - ermin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009959.2 | 681 | Missense Mutation | GAT,GGT | D,G 196 | NP_001009959.1 | |
NM_001304344.1 | 681 | Missense Mutation | GAT,GGT | D,G 183 | NP_001291273.1 | |
NM_001304345.1 | 681 | Missense Mutation | GAT,GGT | D,G 183 | NP_001291274.1 | |
NM_001304346.1 | 681 | Missense Mutation | GAT,GGT | D,G 163 | NP_001291275.1 | |
NM_020711.2 | 681 | Missense Mutation | GAT,GGT | D,G 183 | NP_065762.1 |
GALNT5 - polypeptide N-acetylgalactosaminyltransferase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |