Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGATGACCCCCCGGCGCTGCTTAAC[A/G]AAGTCAAAGGCCTCGTCCAGCCGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608860 MIM: 603068 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASTL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASTL - astacin-like metallo-endopeptidase (M12 family) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002036.3 | 944 | Intron | NP_001002036.3 | |||
XM_011511205.2 | 944 | Intron | XP_011509507.1 | |||
XM_011511206.2 | 944 | Intron | XP_011509508.1 | |||
XM_011511207.2 | 944 | Intron | XP_011509509.1 | |||
XM_011511208.2 | 944 | Intron | XP_011509510.1 |
DUSP2 - dual specificity phosphatase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004418.3 | 944 | Silent Mutation | TTC,TTT | F,F 286 | NP_004409.1 | |
XM_017003546.1 | 944 | Silent Mutation | TTC,TTT | F,F 314 | XP_016859035.1 |