Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGTAGCATGCATACCAATATTTC[A/G]CCATGCAAGCAGTGGCGGAGTCCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609576 | ||||||||||||||||||||
Literature Links: |
ACADL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACADL - acyl-CoA dehydrogenase, long chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001608.3 | 1759 | Missense Mutation | GCG,GTG | A,V 368 | NP_001599.1 | |
XM_005246517.4 | 1759 | Missense Mutation | GCG,GTG | A,V 347 | XP_005246574.1 | |
XM_017003955.1 | 1759 | Missense Mutation | GCG,GTG | A,V 227 | XP_016859444.1 |