Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGCTGCTGCTGCTGCTGCTGCTG[A/G]GCCTGAGGCTACAGCTCTCCCTGGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 171800 | ||||||||||||||||||||
Literature Links: |
ALPP PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALPP - alkaline phosphatase, placental | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001632.4 | 309 | Missense Mutation | AGC,GGC | S,G 14 | NP_001623.3 |
ECEL1P2 - endothelin converting enzyme like 1 pseudogene 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |