Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAGGCCATCCACGGGGTGCAGGA[C/T]GGGGACCAAGGTGATGCGGGGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605207 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP26B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP26B1 - cytochrome P450 family 26 subfamily B member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001277742.1 | 1238 | Missense Mutation | ATC,GTC | I,V 404 | NP_001264671.1 | |
NM_019885.3 | 1238 | Missense Mutation | ATC,GTC | I,V 479 | NP_063938.1 | |
XM_005264433.4 | 1238 | Missense Mutation | ATC,GTC | I,V 421 | XP_005264490.1 | |
XM_011532988.1 | 1238 | Missense Mutation | ATC,GTC | I,V 288 | XP_011531290.1 |