Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATCCATTGGGTACATGATTTTTT[A/C]TGGTAGGCTAATGGGAGAACAAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611035 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APLF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APLF - aprataxin and PNKP like factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FBXO48 - F-box protein 48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024680.2 | 869 | Nonsense Mutation | GAA,TAA | E,* 134 | NP_001019851.1 | |
XM_005264407.2 | 869 | Nonsense Mutation | GAA,TAA | E,* 134 | XP_005264464.1 | |
XM_017004437.1 | 869 | Nonsense Mutation | GAA,TAA | E,* 134 | XP_016859926.1 |