Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGACGAGTGAAAAGAACTTCATAAA[C/T]GAGGAAAAAGACCAACCCGCAGACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605323 MIM: 612152 MIM: 600266 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CTDSP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CTDSP1 - CTD small phosphatase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206878.1 | Intron | NP_001193807.1 | ||||
NM_021198.2 | Intron | NP_067021.1 | ||||
NM_182642.2 | Intron | NP_872580.1 | ||||
XM_011511569.2 | Intron | XP_011509871.1 | ||||
XM_011511570.2 | Intron | XP_011509872.1 | ||||
XM_017004615.1 | Intron | XP_016860104.1 | ||||
XM_017004616.1 | Intron | XP_016860105.1 | ||||
XM_017004617.1 | Intron | XP_016860106.1 | ||||
XM_017004618.1 | Intron | XP_016860107.1 |
MIR26B - microRNA 26b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC11A1 - solute carrier family 11 member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |