Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGAACACAACCACCCACAAGTCA[C/T]TGGGGTAGAAGCGGTCACAGATATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 162643 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CXCR4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CXCR4 - C-X-C motif chemokine receptor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008540.1 | 891 | Missense Mutation | AAT,AGT | N,S 196 | NP_001008540.1 | |
NM_003467.2 | 891 | Missense Mutation | AAT,AGT | N,S 192 | NP_003458.1 |