Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGAAACATTGCCCAAAGAAGAC[A/C]TCATCAAGTTTGCCAAGAAACAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612711 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GCC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GCC2 - GRIP and coiled-coil domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181453.3 | 242 | Missense Mutation | ATC,CTC | I,L 30 | NP_852118.1 | |
XM_006712870.3 | 242 | Missense Mutation | ATC,CTC | I,L 33 | XP_006712933.1 | |
XM_006712871.1 | 242 | Intron | XP_006712934.1 | |||
XM_006712872.3 | 242 | Missense Mutation | ATC,CTC | I,L 33 | XP_006712935.1 | |
XM_011512213.2 | 242 | UTR 5 | XP_011510515.1 |