Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACGCCGAGAGGCTCTGGCCTTAC[A/G]GGGGTCCCTGGAGTTTGATGATGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613734 MIM: 612981 | ||||||||||||||||||||
Literature Links: |
CCDC115 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC115 - coiled-coil domain containing 115 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IMP4 - IMP4 homolog, U3 small nucleolar ribonucleoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320304.1 | 869 | Missense Mutation | CAG,CGG | Q,R 54 | NP_001307233.1 | |
NM_001320305.1 | 869 | Missense Mutation | CAG,CGG | Q,R 54 | NP_001307234.1 | |
NM_001320306.1 | 869 | UTR 5 | NP_001307235.1 | |||
NM_001320307.1 | 869 | UTR 5 | NP_001307236.1 | |||
NM_001320309.1 | 869 | UTR 5 | NP_001307238.1 | |||
NM_001320310.1 | 869 | UTR 5 | NP_001307239.1 | |||
NM_001320311.1 | 869 | UTR 5 | NP_001307240.1 | |||
NM_033416.2 | 869 | Missense Mutation | CAG,CGG | Q,R 54 | NP_219484.1 |